• USA Home
  • 88503 - HEPES Sodium salt pH 7.5; 0.05M – 0.1M Sodium chloride, L-Glutathione red. 0.005M – L-Glutathione ox. 0.001M solution

EMAIL THIS PAGE TO A FRIEND
88503 Sigma-Aldrich

HEPES Sodium salt pH 7.5; 0.05M – 0.1M Sodium chloride, L-Glutathione red. 0.005M – L-Glutathione ox. 0.001M solution

Synonym: Crystallization solution 31/Kit-No 96827

Purchase

Properties

pH   7.5±0.2 ( in neat)
density   1.01 g/mL at 20 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

38185 Quantification kit for oxidized and reduced glutathione, 200 tests

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?