• USA Home
  • 88546 - 1,1′-Thiocarbonyldiimidazole

EMAIL THIS PAGE TO A FRIEND
88546 Sigma-Aldrich

1,1′-Thiocarbonyldiimidazole

technical, ≥90% (S)

  • CAS Number 6160-65-2

  • Empirical Formula (Hill Notation) C7H6N4S

  • Molecular Weight 178.21

  •  Beilstein Registry Number 609349

  •  EC Number 228-183-4

  •  MDL number MFCD00005289

  •  PubChem Substance ID 24889111

Purchase

Properties

Related Categories Carbonyldiimidazole Derivatives, Chemical Synthesis, Coupling, Synthetic Reagents
InChI Key   RAFNCPHFRHZCPS-UHFFFAOYSA-N
grade   technical
assay   ≥90% (S)
mp   98-102 °C(lit.)
  98-102 °C
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

156051 1,1′-Thiocarbonyldiimidazole, technical grade, 90%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?