• USA Home
  • 88625 - S-tert-Butyl thioacetate

EMAIL THIS PAGE TO A FRIEND
88625 Sigma-Aldrich

S-tert-Butyl thioacetate

purum, ≥98.0% (GC)

  • CAS Number 999-90-6

  • Linear Formula CH3COSC(CH3)3

  • Molecular Weight 132.22

  •  Beilstein Registry Number 1740436

  •  MDL number MFCD00042941

  •  PubChem Substance ID 24888975

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiol Esters More...
InChI Key   PJFHXBBYLAVSLJ-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.453(lit.)
  n20/D 1.453
bp   135-136 °C(lit.)
density   0.927 g/mL at 25 °C(lit.)

Description

Other Notes

The lithium enolate of S-t-butyl thioacetate adds to α,β-unsaturated ketones in the β-position

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
93.2 °F
Flash Point(C) 
34 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

416738 S-tert-Butyl thioacetate, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?