EMAIL THIS PAGE TO A FRIEND
88642 Sigma-Aldrich

1-Thioglycerol

dist., ≥98.0% (GC)

Synonym: α-Monothioglycerol, α-Thioglycerol, 3-Mercapto-1,2-propanediol

  • CAS Number 96-27-5

  • Linear Formula HSCH2CH(OH)CH2OH

  • Molecular Weight 108.16

  •  Beilstein Registry Number 1732046

  •  EC Number 202-495-0

  •  MDL number MFCD00004879

  •  PubChem Substance ID 329769666

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiols/Mercaptans More...
InChI Key   PJUIMOJAAPLTRJ-UHFFFAOYSA-N
assay   ≥98.0% (GC)
  ~90% (RT)
quality   dist.
impurities   ~10% water
refractive index   n20/D 1.527(lit.)
bp   118 °C/5 mmHg(lit.)
density   1.25 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2810 6.1 / PGIII
WGK Germany 
3
RTECS 
TY8140000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

M1753 1-Thioglycerol, ≥97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?