EMAIL THIS PAGE TO A FRIEND
88810 Sigma-Aldrich

Thiourea

puriss. p.a., ACS reagent, ≥99.0%

Synonym: Sulfourea, Thiocarbamide

  • CAS Number 62-56-6

  • Linear Formula NH2CSNH2

  • Molecular Weight 76.12

  •  Beilstein Registry Number 605327

  •  EC Number 200-543-5

  •  MDL number MFCD00008067

  •  PubChem Substance ID 24889147

Purchase

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Puriss p.a. ACS, T-Z, Puriss p.a. ACS More...
grade   ACS reagent
InChI Key   UMGDCJDMYOKAJW-UHFFFAOYSA-N
grade   puriss. p.a.
assay   ≥99.0%
ign. residue   ≤0.1% (as SO4)
loss   ≤0.5% loss on drying, 105°C, 2h
mp   170-176 °C(lit.)
  174-177 °C
solubility   water: soluble137 g/L at 20 °C
anion traces   sulfate (SO42-): ≤100 mg/kg
cation traces   Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤50 mg/kg
  Mg: ≤10 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤5 mg/kg
Gene Information   mouse ... Ephx2(13850)

Description

Application

Chaotropic agent; strong denaturant. Increases solubility and recovery of proteins

Thiourea may be used in the synthesis of thiobarbiturates and graphitic carbon nitride (g-C3N4).

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
YU2800000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?