EMAIL THIS PAGE TO A FRIEND
88940 Sigma-Aldrich

Thionyl bromide

purum, ≥97.0% (AT)

Purchase

Properties

Related Categories Bromination, C-X Bond Formation (Halogen), Chemical Synthesis, Synthetic Reagents
vapor pressure   10 mmHg ( 31 °C)
InChI Key   HFRXJVQOXRXOPP-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (AT)
refractive index   n20/D 1.675(lit.)
bp   48 °C/20 mmHg(lit.)
mp   −52 °C(lit.)
density   2.683 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 3264 8 / PGI
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

251259 Thionyl bromide, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?