EMAIL THIS PAGE TO A FRIEND
88952 Sigma-Aldrich

Thionyl chloride

ReagentPlus®, ≥99.0%

  • CAS Number 7719-09-7

  • Linear Formula SOCl2

  • Molecular Weight 118.97

  •  Beilstein Registry Number 1209273

  •  EC Number 231-748-8

  •  MDL number MFCD00011449

Purchase

Properties

Related Categories C-X Bond Formation (Halogen), Chemical Synthesis, Chlorination, Synthetic Reagents
grade   ReagentPlus®
vapor pressure   97 mmHg ( 20 °C)
InChI Key   FYSNRJHAOHDILO-UHFFFAOYSA-N
assay   ≥99.0%
refractive index   n20/D 1.518(lit.)
bp   79 °C(lit.)
mp   −105 °C(lit.)
density   1.64 g/mL at 20 °C
  1.631 g/mL at 25 °C(lit.)

Description

Legal Information

ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Other Notes

This product has been replaced by 230464-Sigma-Aldrich | Thionyl chloride, ReagentPlus, ≥99% | SOCl2

Price and Availability


Palau Chlor
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Contact with water liberates toxic gas., Reacts violently with water.
RIDADR 
UN 1836 8 / PGI
WGK Germany 
1
RTECS 
XM5150000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?