• USA Home
  • 89011 - 2-Thiophenecarbonyl chloride

EMAIL THIS PAGE TO A FRIEND
89011 Sigma-Aldrich

2-Thiophenecarbonyl chloride

technical, ≥97.0% (AT)

Synonym: α-Thenoyl chloride

  • CAS Number 5271-67-0

  • Empirical Formula (Hill Notation) C5H3ClOS

  • Molecular Weight 146.59

  •  Beilstein Registry Number 110145

  •  EC Number 226-092-4

  •  MDL number MFCD00005428

  •  PubChem Substance ID 24889163

Purchase

Properties

Related Categories Building Blocks, C4 to C6, Chemical Synthesis, Heterocyclic Building Blocks, Thiophenes More...
InChI Key   QIQITDHWZYEEPA-UHFFFAOYSA-N
grade   technical
assay   ≥97.0% (AT)
refractive index   n20/D 1.590(lit.)
bp   206-208 °C(lit.)
density   1.371 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGII
WGK Germany 
3
Flash Point(F) 
195.8 °F
Flash Point(C) 
91 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?