EMAIL THIS PAGE TO A FRIEND
89021 Sigma-Aldrich

Thiophenol

purum, ≥98.0% (GC)

Synonym: Benzenethiol, Phenyl mercaptan

  • CAS Number 108-98-5

  • Linear Formula C6H5SH

  • Molecular Weight 110.18

  •  Beilstein Registry Number 506523

  •  EC Number 203-635-3

  •  MDL number MFCD00004826

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiols/Mercaptans More...
vapor density   3.8 (vs air)
vapor pressure   1.4 mmHg ( 20 °C)
InChI Key   RMVRSNDYEFQCLF-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.588(lit.)
bp   169 °C(lit.)
mp   −15 °C(lit.)
density   1.073 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Target organs 
Kidney, Nervous system
RIDADR 
UN 2337 3(6.1) / PGI
WGK Germany 
3
RTECS 
DC0525000
Flash Point(F) 
122 °F
Flash Point(C) 
50 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?