EMAIL THIS PAGE TO A FRIEND
89027 Sigma-Aldrich

Sodium thiophenolate

purum, ≥96.5% (RT)

Synonym: Benzenethiol sodium salt, Sodium thiophenoxide, Thiophenol sodium salt

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiol/Mercaptan Salts More...
InChI Key   RZWQDAUIUBVCDD-UHFFFAOYSA-M
grade   purum
assay   ≥96.5% (RT)
mp   >300 °C(lit.)

Description

Other Notes

Reagent used in aromatic substitution reactions; Demethylation of quaternary ammonium salts

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1759 8 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

418293 Sodium thiophenolate, technical grade, 90%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?