EMAIL THIS PAGE TO A FRIEND
89051 Sigma-Aldrich

Thiosemicarbazide

purum, ≥98.0% (RT)

Synonym: Thiocarbamoyl hydrazide

  • CAS Number 79-19-6

  • Linear Formula NH2CSNHNH2

  • Molecular Weight 91.14

  •  Beilstein Registry Number 506320

  •  EC Number 201-184-7

  •  MDL number MFCD00007620

  •  PubChem Substance ID 24889171

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiocarbonyl Compounds More...
InChI Key   BRWIZMBXBAOCCF-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (RT)
mp   180-183 °C (dec.)(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3
RTECS 
VT4200000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?