• USA Home
  • 89070 - Silica gel on TLC PET-foils

EMAIL THIS PAGE TO A FRIEND
89070 Sigma-Aldrich

Silica gel on TLC PET-foils

silica gel matrix

Purchase

Properties

Related Categories Analytical/Chromatography, LPLC/TLC/Paper Chromatography, Poly TLC Plates: Silica matrix, TLC Plates, PET foil backing, TLC and HPTLC Plates,
material   PET support
  silica gel matrix
feature   binder Proprietary
  fluorescent indicator
packaging   pkg of 50 ea
mfr. no.   Sigma-Aldrich
application(s)   thin layer chromatography (TLC): suitable
L × W   4 cm × 8 cm
layer thickness   250 μm
particle size   8.0-12.0 μm
pore size   60 Å medium pore diameter

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Silica gel on TLC-PET foils was used to perform thin layer chromatography.

General description

Polyethylene terephthalate (PET) is a thermoplastic polymer resin. It is used on TLC plates as it prevents the chemical reagents to diffuse into the polymer matrix. The chemical degradation of PET is not easy. Silica gel is a rigid 3D network of colloidal silica. It is exclusively used as a support material for the active titanium(III) centers in Ziegler-Natta catalysts. Silica gel allows a controlled fragmentation resulting in the formation of uniform polymer particles with narrow particle size distribution and high bulk density. Silica gel is classified as below:
a. Aquagel - pores are filled with water
b. Xerogel - by the process of evaporation, aqueous phase in the pores are removed
c. Aerogel - solvent is removed by supercritical extraction

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?