EMAIL THIS PAGE TO A FRIEND
89173 Sigma-Aldrich

D-Threitol

≥98.0% (sum of enantiomers, TLC)

Synonym: (2R,3R)-1,2,3,4-Butanetetrol

  • CAS Number 2418-52-2

  • Linear Formula HOCH2[CH(OH)]2CH2OH

  • Molecular Weight 122.12

  •  Beilstein Registry Number 1719752

  •  MDL number MFCD00067036

  •  PubChem Substance ID 329769709

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrate Chemistry, Carbohydrate Derivatives, Carbohydrates, Carbohydrates A to Z,
InChI Key   UNXHWFMMPAWVPI-QWWZWVQMSA-N
assay   ≥98.0% (sum of enantiomers, TLC)
optical activity   [α]20/D −14.4±0.5°, c = 2% in ethanol
mp   87-90 °C
  88-90 °C(lit.)

Description

Other Notes

Chiral tetrol; preparation of chiral bis-crown ethers; Synthesis of the threo-L-tetramine via a tetrasulfonate; Preparation of chiral building blocks

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?