EMAIL THIS PAGE TO A FRIEND
89175 Sigma-Aldrich

L-Threitol

≥98.0% (sum of enantiomers, TLC)

Synonym: (2S,3S)-1,2,3,4-Butanetetrol

  • CAS Number 2319-57-5

  • Linear Formula HOCH2[CH(OH)]2CH2OH

  • Molecular Weight 122.12

  •  Beilstein Registry Number 1719754

  •  MDL number MFCD00064294

  •  PubChem Substance ID 24889189

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrate Chemistry, Carbohydrate Derivatives, Carbohydrates, Carbohydrates A to Z,
InChI Key   UNXHWFMMPAWVPI-IMJSIDKUSA-N
assay   ≥98.0% (sum of enantiomers, TLC)
optical activity   [α]20/D +14.5±0.7°, c = 2% in ethanol
mp   87-90 °C(lit.)
  87-90 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?