• USA Home
  • 89276 - 3-(Chlorocarbonyl)-6,7-dimethoxy-1-methyl-2(1H)-quinoxalinone

EMAIL THIS PAGE TO A FRIEND
89276 Sigma-Aldrich

3-(Chlorocarbonyl)-6,7-dimethoxy-1-methyl-2(1H)-quinoxalinone

for HPLC derivatization, ≥95.0% (HPLC)

Synonym: 3,4-Dihydro-6,7-dimethoxy-4-methyl-3-oxo-2-quinoxalinecarbonyl chloride, 6,7-Dimethoxy-1-methyl-2(1H)-quinoxalinone-3-carbonyl chloride, DMEQ-COCl

  • CAS Number 104077-15-8

  • Empirical Formula (Hill Notation) C12H11ClN2O4

  • Molecular Weight 282.68

  •  Beilstein Registry Number 4198803

  •  MDL number MFCD01075069

  •  PubChem Substance ID 329769719

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Carbonyl Chlorides, Derivatization Reagents, Derivatization Reagents HPLC,
grade   for HPLC derivatization
InChI Key   HQUXEWBKFUSDKC-UHFFFAOYSA-N
assay   ≥95.0% (HPLC)
storage temp.   −20°C

Description

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3261 8 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?