EMAIL THIS PAGE TO A FRIEND
89329 Sigma-Aldrich

Thymol

tested according to Ph.Eur.

Synonym: 2-Isopropyl-5-methylphenol, 5-Methyl-2-(1-methylethyl)phenol, 5-Methyl-2-isopropylphenol, IPMP, Thymolum

  • CAS Number 89-83-8

  • Linear Formula 2-[(CH3)2CH]C6H3-5-(CH3)OH

  • Molecular Weight 150.22

  •  Beilstein Registry Number 1907135

  •  EC Number 201-944-8

  •  MDL number MFCD00002309

  •  PubChem Substance ID 329769724

Purchase

Properties

Related Categories Cell Biology, Nutrition Research, Other Biochemical, Phytochemicals by Chemical Classification
vapor pressure   1 mmHg ( 64 °C)
InChI Key   MGSRCZKZVOBKFT-UHFFFAOYSA-N
pharmacopeia   testing conforms to Pharmacopeia
bp   232 °C(lit.)
mp   48-51 °C(lit.)
density   0.965 g/mL at 25 °C(lit.)

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN2430 - class 8 - PG 3 - EHS - Alkylphenols, solid, n.o.s., HI: A,S
WGK Germany 
2
RTECS 
XP2275000
Flash Point(F) 
213.8 °F
Flash Point(C) 
101 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?