EMAIL THIS PAGE TO A FRIEND
89554 Sigma-Aldrich

TNT buffer solution

BioUltra, for molecular biology, pH 8.0

Synonym: TRIS NaCl Tween® 20 buffer solution

Purchase

Properties

Related Categories Alphabetical, BioUltra, BioUltra Buffers, Biochemicals and Reagents, Biological Buffers,
grade   for molecular biology
product line   BioUltra
impurities   DNases, none detected
  RNases, none detected
  insoluble matter, passes filter test
  phosphatases, none detected
  proteases, none detected
pH   8.0±0.2 (25 °C)
cation traces   Al: ≤1 mg/kg
  Ba: ≤1 mg/kg
  Bi: ≤1 mg/kg
  Ca: ≤5 mg/kg
  Cd: ≤1 mg/kg
  Co: ≤1 mg/kg
  Cr: ≤1 mg/kg
  Cu: ≤1 mg/kg
  Fe: ≤1 mg/kg
  K: ≤20 mg/kg
  Li: ≤1 mg/kg
  Mg: ≤1 mg/kg
  Mn: ≤1 mg/kg
  Mo: ≤1 mg/kg
  Ni: ≤1 mg/kg
  Pb: ≤1 mg/kg
  Sr: ≤1 mg/kg
  Zn: ≤1 mg/kg
λ   neat
UV absorption   λ: 260 nm Amax: 0.04
  λ: 280 nm Amax: 0.03

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

TRIS NaCl Tween 20 (TNT) buffer solution, pH 8.0 may be used in molecular biology as a washing buffer and potentially as an elution buffer for immuno micro sensors.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Price and Availability


Interactive Buffer Table
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?