• USA Home
  • 89568 - (−)-α-Methylbenzyl isothiocyanate

EMAIL THIS PAGE TO A FRIEND
89568 Sigma-Aldrich

(−)-α-Methylbenzyl isothiocyanate

for chiral derivatization, ≥99.0%

Synonym: (−)-1-Phenylethyl isothiocyanate

  • CAS Number 24277-44-9

  • Empirical Formula (Hill Notation) C9H9NS

  • Molecular Weight 163.24

  •  Beilstein Registry Number 6476424

  •  MDL number MFCD00135478

  •  PubChem Substance ID 329769748

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Chiral, Derivatization Reagents, Derivatization Reagents HPLC More...
grade   for chiral derivatization
InChI Key   QQCJPTVZIZVKEZ-MRVPVSSYSA-N
assay   ≥99.0% (sum of enantiomers, GC)
  ≥99.0%
optical purity   enantiomeric ratio: ≥97:3 (GC)
color   very deep yellow
storage temp.   2-8°C

Description

Other Notes

Derivatizing agent for HPLC; assay of enantiomeric purity of amines

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
150.8 °F
Flash Point(C) 
66 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?