EMAIL THIS PAGE TO A FRIEND
89670 Sigma-Aldrich

p-Tolunitrile

purum, ≥98.0% (GC)

Synonym: 4-Methylbenzonitrile

  • CAS Number 104-85-8

  • Linear Formula CH3C6H4CN

  • Molecular Weight 117.15

  •  Beilstein Registry Number 507386

  •  EC Number 203-244-8

  •  MDL number MFCD00001827

Purchase

Properties

Related Categories Building Blocks, C8 to C9, Chemical Synthesis, Cyanides/Nitriles, Nitrogen Compounds,
InChI Key   VCZNNAKNUVJVGX-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
bp   103-106 °C/20 mmHg(lit.)
  217 °C(lit.)
mp   26-28 °C(lit.)
density   0.981 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
XV0700000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?