EMAIL THIS PAGE TO A FRIEND
89676 Sigma-Aldrich

Toluene

for residue analysis, ≥99.8% (GC)

  • CAS Number 108-88-3

  • Linear Formula C6H5CH3

  • Molecular Weight 92.14

  •  Beilstein Registry Number 635760

  •  EC Number 203-625-9

  •  MDL number MFCD00008512

Purchase

Properties

grade   for residue analysis
vapor density   3.2 (vs air)
vapor pressure   22 mmHg ( 20 °C)
  26 mmHg ( 25 °C)
InChI Key   YXFVVABEGXRONW-UHFFFAOYSA-N
assay   ≥99.8% (GC)
autoignition temp.   997 °F
expl. lim.   7 %
impurities   ≤0.02% water
evapn. residue   ≤0.0005%
refractive index   n/D 1.496(lit.)
  n20/D 1.497
bp   110-111 °C(lit.)
mp   -93 °C(lit.)
density   0.865 g/mL at 25 °C(lit.)

Description

General description

no impurities present which would show greater signals than 5 ng/l lindane, in the GC/ECD retention time range of lindane to DDT

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1294 - class 3 - PG 2 - Toluene
WGK Germany 
2
RTECS 
XS5250000
Flash Point(F) 
39.2 °F
Flash Point(C) 
4 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?