• USA Home
  • 89715 - (−)-(1R)-Menthyl (S)-p-toluenesulfinate

EMAIL THIS PAGE TO A FRIEND
89715 Sigma-Aldrich

(−)-(1R)-Menthyl (S)-p-toluenesulfinate

purum, ≥98.0% (sum of enantiomers, HPLC)

  • CAS Number 1517-82-4

  • Empirical Formula (Hill Notation) C17H26O2S

  • Molecular Weight 294.45

  •  Beilstein Registry Number 3207468

  •  MDL number MFCD00010192

  •  PubChem Substance ID 329769528

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks, Sulfonates/Sulfinates More...
InChI Key   NQICGNSARVCSGJ-BFNFPZTMSA-N
grade   purum
assay   ≥98.0% (sum of enantiomers, HPLC)
optical activity   [α]20/D −197±7°, c = 2% in acetone
mp   99-104 °C
storage temp.   −20°C

Description

Other Notes

Reagent for the preparation of optically active sulfoxides by Grignard-displacement of the O-menthyl group; employed as chiral reagents

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?