• USA Home
  • 89775 - p-Toluenesulfonic anhydride

EMAIL THIS PAGE TO A FRIEND
89775 Sigma-Aldrich

p-Toluenesulfonic anhydride

purum, ≥95.0% (NT)

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonic/Sulfinic Acid Anhydrides, Sulfur Compounds More...
InChI Key   PDVFSPNIEOYOQL-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (NT)
mp   121-127 °C(lit.)
  122-132 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2585PSN2 8 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?