• USA Home
  • 89792 - p-Toluenesulfonhydrazide

EMAIL THIS PAGE TO A FRIEND
89792 Sigma-Aldrich

p-Toluenesulfonhydrazide

technical, ≥95% (HPLC)

Synonym: p-Toluenesulfonic acid hydrazide, p-Toluenesulfonyl hydrazide, Tosylhydrazide

  • CAS Number 1576-35-8

  • Linear Formula CH3C6H4SO2NHNH2

  • Molecular Weight 186.23

  •  Beilstein Registry Number 610130

  •  EC Number 216-407-3

  •  MDL number MFCD00007588

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonyl Hydrazides, Sulfur Compounds More...
InChI Key   ICGLPKIVTVWCFT-UHFFFAOYSA-N
grade   technical
assay   ≥95% (HPLC)
mp   103-108 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3226A3 4.1
WGK Germany 
3
RTECS 
MW0210000
Flash Point(F) 
140 °F
Flash Point(C) 
60 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?