EMAIL THIS PAGE TO A FRIEND
89820 Sigma-Aldrich

p-Toluoyl chloride

purum, ≥98.0% (GC)

Synonym: 4-Methylbenzoyl chloride

  • CAS Number 874-60-2

  • Linear Formula CH3C6H4COCl

  • Molecular Weight 154.59

  •  Beilstein Registry Number 471492

  •  EC Number 212-864-8

  •  MDL number MFCD00000696

  •  PubChem Substance ID 24889280

Purchase

Properties

Related Categories Acid Halides, Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks More...
vapor density   5.33 (vs air)
vapor pressure   0.55 mmHg ( 20 °C)
InChI Key   NQUVCRCCRXRJCK-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
expl. lim.   1.6 %, 117 °F
refractive index   n20/D 1.553(lit.)
  n20/D 1.555
bp   225-227 °C(lit.)
  95-96 °C/10 mmHg(lit.)
mp   −4-−2 °C(lit.)
density   1.169 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGII
WGK Germany 
3
RTECS 
XV1660000
Flash Point(F) 
179.6 °F
Flash Point(C) 
82 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?