EMAIL THIS PAGE TO A FRIEND
90030 Sigma-Aldrich

p-Tolylacetic acid

purum, ≥98.0% (T)

Synonym: 4-Methylphenylacetic acid

  • CAS Number 622-47-9

  • Linear Formula CH3C6H4CH2CO2H

  • Molecular Weight 150.17

  •  Beilstein Registry Number 2043528

  •  EC Number 210-738-7

  •  MDL number MFCD00004353

  •  PubChem Substance ID 329769800

Purchase

Properties

Related Categories Building Blocks, C9, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   GXXXUZIRGXYDFP-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (T)
bp   265-267 °C(lit.)
mp   88-92 °C(lit.)
  88-92 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
AJ7569000
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

T38105 p-Tolylacetic acid, ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?