• USA Home
  • 90110 - m-Tolylhydrazine hydrochloride

EMAIL THIS PAGE TO A FRIEND
90110 Sigma-Aldrich

m-Tolylhydrazine hydrochloride

purum, ≥98.0% (AT)

Synonym: 3-Methylphenylhydrazine hydrochloride

  • CAS Number 637-04-7

  • Linear Formula CH3C6H4NHNH2 · HCl

  • Molecular Weight 158.63

  •  Beilstein Registry Number 3563995

  •  EC Number 211-276-9

  •  MDL number MFCD00012932

  •  PubChem Substance ID 24889302

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Hydrazines, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   SHTBTGXNVWYODU-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (AT)
ign. residue   ≤0.5%
mp   184-194 °C (dec.)(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?