• USA Home
  • 90120 - p-Tolylhydrazine hydrochloride

EMAIL THIS PAGE TO A FRIEND
90120 Sigma-Aldrich

p-Tolylhydrazine hydrochloride

purum, ≥98.0% (AT)

Synonym: 4-Methylphenylhydrazine hydrochloride

  • CAS Number 637-60-5

  • Linear Formula CH3C6H4NHNH2 · HCl

  • Molecular Weight 158.63

  •  Beilstein Registry Number 3563996

  •  EC Number 211-295-2

  •  MDL number MFCD00012940

  •  PubChem Substance ID 329769851

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Hydrazines, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   HMHWNJGOHUYVMD-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (AT)
mp   >200 °C (dec.)(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
MW0195000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?