EMAIL THIS PAGE TO A FRIEND
90187 Sigma-Aldrich

T-2 Toxin

≥98.0% (TLC)

Synonym: 12,13-Epoxytrichothec-9-ene-3,4,8,15-tetrol-4,15-diacetate-8-isovalerate, 4β,15-Diacetoxy-3α-hydroxy-8α-(3-methylbutyryloxy)-12,13-epoxytrichothec-9-ene

  • CAS Number 21259-20-1

  • Empirical Formula (Hill Notation) C24H34O9

  • Molecular Weight 466.52

  •  Beilstein Registry Number 3575695

  •  EC Number 244-297-7

  •  MDL number MFCD00871255

  •  PubChem Substance ID 329769858

Purchase

Properties

InChI Key   BXFOFFBJRFZBQZ-QYWOHJEZSA-N
assay   ≥98.0% (TLC)

Description

Caution

mycotoxin; severe poison

Other Notes

Reference in the quantitative determination of trichothecenes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN3462 - class 6.1 - PG 1 - Toxins, extracted from living sources, solid, n.o.s., HI: all (not BR)
WGK Germany 
3
RTECS 
YD0100000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?