• USA Home
  • 90251 - N,N′,N′′-Triacetylchitotriose

EMAIL THIS PAGE TO A FRIEND
90251 Sigma-Aldrich

N,N′,N′′-Triacetylchitotriose

~95% (HPLC)

Synonym: O-[2-Acetamido-2-deoxy-β-D-glucopyranosyl-(1→4)]-O-[2-acetamido-2-deoxy-β-D-glucopyranosyl-(1→4)]-2-acetamido-2-deoxy-D-glucopyranose

  • CAS Number 38864-21-0

  • Empirical Formula (Hill Notation) C24H41N3O16

  • Molecular Weight 627.59

  •  Beilstein Registry Number 75249

  •  MDL number MFCD00136047

  •  PubChem Substance ID 329769894

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Oligosaccharide More...
InChI Key   LRDDKCYRFNJZBX-WHFMPQCRSA-N
assay   ~95% (HPLC)
impurities   ≤6% water
solubility   H2O: 50 mg/mL clear to slightly hazy, colorless to faintly yellow
storage temp.   −20°C

Description

Other Notes

Studies on a chitooligosaccharide-specific lectin

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

T2144 N,N′,N′′-Triacetylchitotriose, ≥93% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?