• USA Home
  • 90461 - Triethylenetetramine dihydrochloride

EMAIL THIS PAGE TO A FRIEND
90461 Sigma-Aldrich

Triethylenetetramine dihydrochloride

puriss., 2×cryst., ≥99.0% (AT)

Synonym: N,N′-Bis(2-aminoethyl)-1,2-ethanediamine dihydrochloride

  • CAS Number 38260-01-4

  • Linear Formula NH2CH2CH2(NHCH2CH2)2NH2 · 2HCl

  • Molecular Weight 219.16

  •  Beilstein Registry Number 5961763

  •  EC Number 253-854-3

  •  MDL number MFCD00012889

  •  PubChem Substance ID 329769909

Purchase

Properties

Related Categories Amine Salts, Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   WYHIICXRPHEJKI-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (AT)
quality   2×cryst.
mp   120-123 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
KH8587085

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

T5033 Triethylenetetramine dihydrochloride

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?