• USA Home
  • 90470 - Triethylenetetramine disulfate salt dihydrate

EMAIL THIS PAGE TO A FRIEND
90470 Sigma-Aldrich

Triethylenetetramine disulfate salt dihydrate

for spectrophotometric det. of Cu, ≥98.0%

  • CAS Number 438209-51-9

  • Linear Formula NH2CH2CH2(NHCH2CH2)2NH2 · 2H2SO4 · 2H2O

  • Molecular Weight 378.42

  •  Beilstein Registry Number 3723212

  •  MDL number MFCD10567441

  •  PubChem Substance ID 329769911

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Spectroscopy, UV/Vis Reagents, UV/Visible (UV/VIS) Spectroscopy More...
InChI Key   FZYPUWMRSXBMOB-UHFFFAOYSA-N
assay   ≥98.0% (T)
  ≥98.0%
quality   for spectrophotometric det. of Cu
ign. residue   ≤0.05%
mp   300-305 °C (dec.)
  300-305 °C(lit.)

Price and Availability


Laboratory Gloves

Spectroquant Prove
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?