• USA Home
  • 90603 - 1,5,7-Triazabicyclo[4.4.0]dec-5-ene bound to polystyrene

EMAIL THIS PAGE TO A FRIEND
90603 Sigma-Aldrich

1,5,7-Triazabicyclo[4.4.0]dec-5-ene bound to polystyrene

extent of labeling: ~2.6 mmol/g base loading

Synonym: 1,3,4,6,7,8-Hexahydro-2H-pyrimido[1,2-a]pyrimidine bound to polystyrene, 7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene on polystyrene

Purchase

Properties

Related Categories Acids & Bases, Bases, Chemical Synthesis, Organic Bases, Solid Supported Synthesis,
extent of labeling   ~2.6 mmol/g base loading
matrix   crosslinked with 2% DVB
particle size   200-400 mesh

Description

Application

Polymer-supported, strong, hindered pentasubstituted guanidine base

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3263 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Related Products

replaced by

Product #

Description

Add to Cart

358754 1,3,4,6,7,8-Hexahydro-2H-pyrimido[1,2-a]pyrimidine, polymer-bound, 200-400 mesh, extent of labeling: 2.6 mmol/g loading, 2 % cross-linked with divinylbenzene

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?