• USA Home
  • 90715 - 1,3,5-Tribromobenzene

EMAIL THIS PAGE TO A FRIEND
90715 Sigma-Aldrich

1,3,5-Tribromobenzene

purum, ≥98.0% (AT)

  • CAS Number 626-39-1

  • Empirical Formula (Hill Notation) C6H3Br3

  • Molecular Weight 314.80

  •  Beilstein Registry Number 1858917

  •  EC Number 210-947-3

  •  MDL number MFCD00000080

  •  PubChem Substance ID 24889400

Purchase

Properties

Related Categories Aryl, Building Blocks, C6, Chemical Synthesis, Halogenated Hydrocarbons,
InChI Key   YWDUZLFWHVQCHY-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (AT)
bp   271 °C(lit.)
mp   117-121 °C(lit.)
  117-121 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?