EMAIL THIS PAGE TO A FRIEND
90915 Sigma-Aldrich

Tributyltin hydride

purum, ≥96.0% (GC)

  • CAS Number 688-73-3

  • Linear Formula [CH3(CH2)3]3SnH

  • Molecular Weight 291.06

  •  Beilstein Registry Number 3587329

  •  EC Number 211-704-4

  •  MDL number MFCD00009416

  •  PubChem Substance ID 329769967

Purchase

Properties

Related Categories Chemical Synthesis, Organometallic Reagents, Organotin, Organotins, Reduction,
InChI Key   DBGVGMSCBYYSLD-UHFFFAOYSA-N
grade   purum
assay   ≥96.0% (GC)
refractive index   n20/D 1.473(lit.)
bp   80 °C/0.4 mmHg(lit.)
density   1.082 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Reducing agent acting via radical intermediates

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

234788 Tributyltin hydride, contains 0.05% BHT as stabilizer, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?