EMAIL THIS PAGE TO A FRIEND
91012 Sigma-Aldrich

Glyceryl tributyrate

technical, ≥94.0% (GC)

Synonym: 1,2,3-Tributyrylglycerol, Glycerol tributyrate, Tributyrin

  • CAS Number 60-01-5

  • Linear Formula (CH3CH2CH2COOCH2)2CHOCOCH2CH2CH3

  • Molecular Weight 302.36

  •  Beilstein Registry Number 1714746

  •  EC Number 200-451-5

  •  MDL number MFCD00009392

  •  PubChem Substance ID 329769991

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, C12 to C63, Carbonyl Compounds, Chemical Synthesis,
InChI Key   UYXTWWCETRIEDR-UHFFFAOYSA-N
grade   technical
assay   ≥94.0% (GC)
refractive index   n20/D 1.435(lit.)
  n20/D 1.435
bp   129-131 °C/0.03 mmHg(lit.)
  287-288 °C(lit.)
density   1.032 g/mL at 20 °C(lit.)

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
ET7350000
Flash Point(F) 
345.2 °F
Flash Point(C) 
174 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?