• USA Home
  • 91029 - Glycerol trihexanoate

EMAIL THIS PAGE TO A FRIEND
91029 Sigma-Aldrich

Glycerol trihexanoate

puriss., ≥99.0% (GC)

Synonym: 1,2,3-Tricaproylglycerol, Glycerol tricapronate, Tricaproin, Trihexanoin

  • CAS Number 621-70-5

  • Linear Formula [CH3(CH2)4COOCH2]2CHOCO(CH2)4CH3

  • Molecular Weight 386.52

  •  Beilstein Registry Number 1806732

  •  EC Number 210-701-5

  •  MDL number MFCD00042905

  •  PubChem Substance ID 329769994

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, C12 to C63, Carbonyl Compounds, Chemical Synthesis,
InChI Key   MAYCICSNZYXLHB-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (GC)
refractive index   n20/D 1.442
density   0.98 g/mL(lit.)

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
MB2700000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

T0888 Glycerol trihexanoate, ~99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?