• USA Home
  • 91120 - 2,2,2-Trichloroethanol

EMAIL THIS PAGE TO A FRIEND
91120 Sigma-Aldrich

2,2,2-Trichloroethanol

purum, ≥98.0% (GC)

Synonym: Trichloroethyl alcohol

  • CAS Number 115-20-8

  • Linear Formula Cl3CCH2OH

  • Molecular Weight 149.40

  •  Beilstein Registry Number 1697495

  •  EC Number 204-071-0

  •  MDL number MFCD00004677

  •  PubChem Substance ID 24889448

Purchase

Properties

Related Categories Alcohols, Building Blocks, C2 to C6, Chemical Synthesis, Organic Building Blocks,
vapor density   5.16 (vs air)
vapor pressure   1 mmHg ( 20 °C)
InChI Key   KPWDGTGXUYRARH-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.49(lit.)
  n20/D 1.491
bp   152-154 °C(lit.)
mp   17.8 °C(lit.)
density   1.56 g/mL at 20 °C(lit.)

Description

Caution

may discolor to brown on storage

Other Notes

A hydrophobic quencher of protein fluorescence, senses the presence of a hydrophobic domain in the vicinity of the tryptophan residues

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
KM3850000
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?