• USA Home
  • 91250 - Ethyl trichloroacetate

EMAIL THIS PAGE TO A FRIEND
91250 Sigma-Aldrich

Ethyl trichloroacetate

purum, ≥96.5% (GC)

  • CAS Number 515-84-4

  • Linear Formula Cl3CCO2CH2CH3

  • Molecular Weight 191.44

  •  Beilstein Registry Number 1761567

  •  EC Number 208-212-7

  •  MDL number MFCD00000795

  •  PubChem Substance ID 329770014

Purchase

Properties

Related Categories Building Blocks, C2 to C5, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   SJMLNDPIJZBEKY-UHFFFAOYSA-N
grade   purum
assay   ≥96.5% (GC)
impurities   ≤3.5% trichloroacetic acid
refractive index   n20/D 1.453(lit.)
  n20/D 1.453
bp   168 °C(lit.)
density   1.378 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
1
Flash Point(F) 
163.4 °F
Flash Point(C) 
73 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?