• USA Home
  • 91340 - 2,4,6-Trichlorophenol

EMAIL THIS PAGE TO A FRIEND
91340 Sigma-Aldrich

2,4,6-Trichlorophenol

purum, ≥97.0% (GC), white to slightly brown

  • CAS Number 88-06-2

  • Linear Formula Cl3C6H2OH

  • Molecular Weight 197.45

  •  Beilstein Registry Number 776729

  •  EC Number 201-795-9

  •  MDL number MFCD00002172

Purchase

Properties

Related Categories Building Blocks, C6 to C8, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds,
vapor pressure   1 mmHg ( 76.5 °C)
InChI Key   LINPIYWFGCPVIE-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
color   white to slightly brown
bp   246 °C(lit.)
mp   64-66 °C(lit.)
  65-68 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Physical form

solidified mass or chunks

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2020 6.1 / PGIII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?