EMAIL THIS PAGE TO A FRIEND
91520 Sigma-Aldrich

1-Tridecanol

purum, ≥98.0% (GC)

Synonym: Tridecyl alcohol

  • CAS Number 112-70-9

  • Linear Formula CH3(CH2)12OH

  • Molecular Weight 200.36

  •  Beilstein Registry Number 1739991

  •  EC Number 203-998-8

  •  MDL number MFCD00004756

  •  PubChem Substance ID 329770048

Purchase

Properties

Related Categories Alcohols, Biochemicals and Reagents, Building Blocks, C11 to C30+, Chemical Synthesis,
vapor density   6.9 (vs air)
InChI Key   XFRVVPUIAFSTFO-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
bp   155-156 °C/15 mmHg(lit.)
mp   29-34 °C(lit.)
  31-34 °C
solubility   chloroform: soluble0.1 g/mL, clear
density   0.822 g/mL at 25 °C(lit.)

Price and Availability


Laboratory Gloves

The Future of Bioimaging
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
YD4200000
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?