EMAIL THIS PAGE TO A FRIEND
91598 Sigma-Aldrich

1-Phenyltridecane

analytical standard

Synonym: Tridecylbenzene

  • CAS Number 123-02-4

  • Linear Formula C6H5(CH2)12CH3

  • Molecular Weight 260.46

  •  Beilstein Registry Number 1947201

  •  EC Number 204-592-3

  •  MDL number MFCD00008980

  •  PubChem Substance ID 24889500

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Aromatics,
grade   analytical standard
InChI Key   MCVUKOYZUCWLQQ-UHFFFAOYSA-N
assay   ≥99.5% (GC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.482(lit.)
  n20/D 1.482
mp   10 °C(lit.)
density   0.881 g/mL at 25 °C(lit.)
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
Flash Point(F) 
>233.6 °F
Flash Point(C) 
>112 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?