• USA Home
  • 91662 - Tridodecylmethylammonium chloride

EMAIL THIS PAGE TO A FRIEND
91662 Sigma-Aldrich

Tridodecylmethylammonium chloride

purum, ≥97.0% (AT)

Synonym: Methyltridodecylammonium chloride

  • CAS Number 7173-54-8

  • Linear Formula [CH3(CH2)11]3N(Cl)CH3

  • Molecular Weight 572.47

  •  Beilstein Registry Number 6031445

  •  MDL number MFCD00043303

  •  PubChem Substance ID 329770064

Purchase

Properties

Related Categories Ammonium Salts, Biochemicals and Reagents, Cationic Detergents, Chemical Synthesis, Detergents,
description   cationic
InChI Key   SBHRWOBHKASWGU-UHFFFAOYSA-M
grade   purum
assay   ≥97.0% (AT)
mp   110-112 °C(lit.)

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?