• USA Home
  • 91720 - Trifluoroacetic anhydride

EMAIL THIS PAGE TO A FRIEND
91720 Sigma-Aldrich

Trifluoroacetic anhydride

purum, ≥99.0% (GC)

Synonym: TFAA

Purchase

Properties

Related Categories Building Blocks, Carbonyl Compounds, Carboxylic Acid Anhydrides, Chemical Synthesis, Organic Building Blocks More...
vapor pressure   6.28 psi ( 20 °C)
InChI Key   QAEDZJGFFMLHHQ-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
refractive index   n20/D 1.3(lit.)
bp   39.5-40 °C(lit.)
mp   −65 °C(lit.)
density   1.511 g/mL at 20 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Powerful activator in the DMSO oxidation of alcohols to carbonyl compounds

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 3265 8 / PGI
WGK Germany 
2
RTECS 
AJ9800000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?