• USA Home
  • 91734 - Ethyl trifluoromethanesulfonate

EMAIL THIS PAGE TO A FRIEND
91734 Sigma-Aldrich

Ethyl trifluoromethanesulfonate

for GC derivatization, ≥99.0%

  • CAS Number 425-75-2

  • Linear Formula CF3SO2OC2H5

  • Molecular Weight 178.13

  •  Beilstein Registry Number 1770746

  •  EC Number 207-037-3

  •  MDL number MFCD00000410

  •  PubChem Substance ID 24889543

Purchase

Properties

Related Categories Acylation Reagents, Analytical Reagents, Analytical/Chromatography, Derivatization Reagents, Derivatization Reagents GC,
grade   for GC derivatization
InChI Key   UVECLJDRPFNRRQ-UHFFFAOYSA-N
assay   ≥99.0% (GC)
  ≥99.0%
refractive index   n20/D 1.336(lit.)
  n20/D 1.337
bp   115 °C(lit.)
density   1.374 g/mL at 25 °C(lit.)

Description

Other Notes

Powerful ethylating agent; review

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2920 3(8) / PGII
WGK Germany 
2
Flash Point(F) 
95 °F
Flash Point(C) 
35 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?