• USA Home
  • 91738 - Trifluoromethanesulfonic acid

EMAIL THIS PAGE TO A FRIEND
91738 Sigma-Aldrich

Trifluoromethanesulfonic acid

purum, ≥98.0% (T)

Synonym: Triflic acid

  • CAS Number 1493-13-6

  • Linear Formula CF3SO3H

  • Molecular Weight 150.08

  •  Beilstein Registry Number 1812100

  •  EC Number 216-087-5

  •  MDL number MFCD00007514

Purchase

Properties

Related Categories Acids, Acids & Bases, Chemical Synthesis, Organic Acids, Synthetic Reagents More...
vapor density   5.2 (vs air)
vapor pressure   8 mmHg ( 25 °C)
InChI Key   ITMCEJHCFYSIIV-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (T)
color   slightly brown
refractive index   n20/D 1.327(lit.)
bp   162 °C(lit.)
density   1.696 g/mL at 25 °C(lit.)

Description

Application

Deglycosylation agent

Other Notes

Review; Reagent for the deglycosylation of glycoproteins

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGII
WGK Germany 
1
Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

158534 Trifluoromethanesulfonic acid, reagent grade, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?