• USA Home
  • 91765 - 4-(Trifluoromethyl)benzaldehyde

EMAIL THIS PAGE TO A FRIEND
91765 Sigma-Aldrich

4-(Trifluoromethyl)benzaldehyde

purum, ≥97.0% (GC)

Synonym: α,α,α-Trifluoro-p-tolualdehyde

  • CAS Number 455-19-6

  • Linear Formula CF3C6H4CHO

  • Molecular Weight 174.12

  •  Beilstein Registry Number 1101680

  •  EC Number 207-240-7

  •  MDL number MFCD00006952

  •  PubChem Substance ID 24889559

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C8, Carbonyl Compounds, Chemical Synthesis,
InChI Key   BEOBZEOPTQQELP-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.463(lit.)
  n20/D 1.464
bp   66-67 °C/13 mmHg(lit.)
density   1.275 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
149 °F
Flash Point(C) 
65 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?