• USA Home
  • 91868 - 3-(Trifluoromethyl)phenylacetic acid

EMAIL THIS PAGE TO A FRIEND
91868 Sigma-Aldrich

3-(Trifluoromethyl)phenylacetic acid

purum, ≥99.0% (T)

Synonym: (α,α,α-Trifluoro-m-tolyl)acetic acid

  • CAS Number 351-35-9

  • Linear Formula CF3C6H4CH2CO2H

  • Molecular Weight 204.15

  •  Beilstein Registry Number 2213223

  •  EC Number 206-511-7

  •  MDL number MFCD00004339

  •  PubChem Substance ID 329770091

Purchase

Properties

Related Categories Building Blocks, C9, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   BLXXCCIBGGBDHI-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (T)
mp   ~75 °C
  76-79 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?