• USA Home
  • 91873 - Trimethyl(trifluoromethyl)silane

EMAIL THIS PAGE TO A FRIEND
91873 Sigma-Aldrich

Trimethyl(trifluoromethyl)silane

purum, ≥98.0% (GC)

Synonym: (Trifluoromethyl)trimethylsilane, Ruppert’s Reagent, TFMTMS

  • CAS Number 81290-20-2

  • Linear Formula (CH3)3SiCF3

  • Molecular Weight 142.19

  •  Beilstein Registry Number 4241868

  •  MDL number MFCD00145454

  •  PubChem Substance ID 329770092

Purchase

Properties

Related Categories C-C Bond Formation, Chemical Synthesis, Other Synthetic Reagents for C-C Bond Formation, Synthetic Reagents
InChI Key   MWKJTNBSKNUMFN-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.332
bp   54-55 °C(lit.)
density   0.962 g/mL at 20 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Reagent for the nucleophilic trifluoromethylation of various compounds induced by F-

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993BF 3 / PGII
WGK Germany 
3
Flash Point(F) 
1.4 °F
Flash Point(C) 
-17 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

488712 Trimethyl(trifluoromethyl)silane, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?