• USA Home
  • 91890 - Trigonelline hydrochloride

EMAIL THIS PAGE TO A FRIEND
91890 Sigma-Aldrich

Trigonelline hydrochloride

≥98.0% (AT)

Synonym: 1-Methylpyridinium-3-carboxylate hydrochloride, N-Methylnicotinic acid betaine hydrochloride

  • CAS Number 6138-41-6

  • Empirical Formula (Hill Notation) C7H7NO2 · HCl

  • Molecular Weight 173.60

  •  Beilstein Registry Number 3734169

  •  EC Number 228-119-5

  •  MDL number MFCD00077250

  •  PubChem Substance ID 329770093

Purchase

Properties

Related Categories Building Blocks, C7, Chemical Synthesis, Heterocyclic Building Blocks, Pyridines More...
InChI Key   TZSYLWAXZMNUJB-UHFFFAOYSA-N
assay   ≥98.0% (AT)
form   powder or crystals
mp   ~260 °C (dec.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?