EMAIL THIS PAGE TO A FRIEND
91905 Sigma-Aldrich

Trihexylamine

purum, ≥97.0% (GC)

  • CAS Number 102-86-3

  • Linear Formula [CH3(CH2)5]3N

  • Molecular Weight 269.51

  •  Beilstein Registry Number 1755828

  •  EC Number 203-062-9

  •  MDL number MFCD00009523

  •  PubChem Substance ID 329770096

Purchase

Properties

vapor density   9.3 (vs air)
vapor pressure   <1 mmHg ( 20 °C)
InChI Key   DIAIBWNEUYXDNL-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.442(lit.)
  n20/D 1.442
bp   150-159 °C/12 mmHg(lit.)
  263-265 °C(lit.)
density   0.794 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?